Assay Details
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630681.3 | XP_005630738.1 | 7 - 8 | 1160 | 114 | |
XM_005630682.3 | XP_005630739.1 | 6 - 7 | 1002 | 114 | ||
XM_014120701.2 | XP_013976176.1 | 6 - 7 | 998 | 114 | ||
XM_533023.6 | XP_533023.3 | 7 - 8 | 1155 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
