Assay Details
Gene Symbol: MLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627487.3 | XP_005627544.1 | 13 - 14 | 2962 | 74 | |
XM_005627488.3 | XP_005627545.1 | 13 - 14 | 2954 | 74 | ||
XM_005627489.3 | XP_005627546.1 | 11 - 12 | 3862 | 74 | ||
XM_005627490.3 | XP_005627547.1 | 12 - 13 | 2757 | 74 | ||
XM_014118057.2 | XP_013973532.1 | 13 - 14 | 2884 | 74 | ||
XM_014118058.2 | XP_013973533.1 | 12 - 13 | 2878 | 74 | ||
XM_014118059.2 | XP_013973534.1 | 12 - 13 | 2806 | 74 | ||
XM_014118060.2 | XP_013973535.1 | 13 - 14 | 2962 | 74 | ||
XM_014118061.2 | XP_013973536.1 | 13 - 14 | 2962 | 74 | ||
XM_014118063.2 | XP_013973538.1 | 12 - 13 | 1398 | 74 | ||
XM_022426160.1 | XP_022281868.1 | 10 - 11 | 2627 | 74 | ||
XM_022426161.1 | XP_022281869.1 | 10 - 11 | 2788 | 74 | ||
XM_022426162.1 | XP_022281870.1 | 11 - 12 | 2620 | 74 | ||
XM_022426163.1 | XP_022281871.1 | 9 - 10 | 2398 | 74 | ||
XM_022426164.1 | XP_022281872.1 | 9 - 10 | 2403 | 74 | ||
XM_022426165.1 | XP_022281873.1 | 9 - 10 | 2588 | 74 | ||
XM_532178.6 | XP_532178.4 | 11 - 12 | 2690 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
