Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_533002.5 | XP_533002.2 | 6 - 7 | 775 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
