Assay Details
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284449.1 | NP_001271378.1 | 2 - 3 | 175 | 76 | |
GenBank mRNA | AB793325.1 | - | 2 - 3 | 176 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
