Assay Details
Gene Symbol: PSENEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284449.1 | NP_001271378.1 | 3 - 4 | 289 | 75 | |
GenBank mRNA | AB793325.1 | - | 3 - 4 | 290 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
