Assay Details
Gene Symbol: RPS6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252170.1 | NP_001239099.1 | 3 - 4 | 416 | 79 | |
GenBank mRNA | GR893301.1 | - | 3 - 4 | 416 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
