Assay Details
Gene Symbol: MFSD4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_536102.6 | XP_536102.3 | 4 - 5 | 940 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
