Assay Details
Gene Symbol: JHY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619653.3 | XP_005619710.1 | 104 | 94 | ||
XM_022418180.1 | XP_022273888.1 | 105 | 94 | |||
XM_536544.6 | XP_536544.2 | 104 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005619653.3, XM_022418180.1, XM_536544.6 transcripts.
Set Membership: |
![]() ![]() |
