Assay Details
Gene Symbol: ETNK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014111180.2 | XP_013966655.1 | 3 - 4 | 548 | 88 | |
XM_022415390.1 | XP_022271098.1 | 2 - 3 | 428 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
