Assay Details
Gene Symbol: MAPK1IP1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623420.3 | XP_005623477.1 | 3 - 4 | 751 | 85 | |
XM_005623421.2 | XP_005623478.1 | 3 - 4 | 752 | 85 | ||
XM_022422023.1 | XP_022277731.1 | 3 - 4 | 750 | 85 | ||
XM_022422024.1 | XP_022277732.1 | 3 - 4 | 952 | 85 | ||
XM_022422025.1 | XP_022277733.1 | 2 - 3 | 848 | 85 | ||
XM_537452.5 | XP_537452.2 | 3 - 4 | 870 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
