Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417737.1 | XP_022273445.1 | 2 - 3 | 194 | 76 | |
XM_536445.5 | XP_536445.2 | 2 - 3 | 232 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
