Assay Details
Gene Symbol: PTTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022417737.1 | XP_022273445.1 | 3 - 4 | 378 | 80 | |
XM_022417738.1 | XP_022273446.1 | 3 - 4 | 413 | 80 | ||
XM_536445.5 | XP_536445.2 | 3 - 4 | 416 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
