Assay Details
Gene Symbol: LTB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033510.1 | NP_001028682.1 | 3 - 4 | 290 | 109 | |
XM_022425731.1 | XP_022281439.1 | 4 - 5 | 765 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
