Assay Details
Gene Symbol: IQCF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632477.2 | XP_005632534.1 | 1 - 2 | 156 | 76 | |
XM_541864.6 | XP_541864.2 | 1 - 2 | 158 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005632477.2, XM_541864.6 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
