Assay Details
Gene Symbol: RWDD2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005627582.2 | XP_005627639.1 | 1 - 2 | 1005 | 72 | |
XM_005627583.3 | XP_005627640.1 | 2 - 3 | 526 | 72 | ||
XM_539022.5 | XP_539022.1 | 2 - 3 | 483 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
