Assay Details
Gene Symbol: ELP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014121289.2 | XP_013976764.1 | 7 - 8 | 972 | 144 | |
XM_022405381.1 | XP_022261089.1 | 7 - 8 | 972 | 144 | ||
XM_540560.6 | XP_540560.2 | 7 - 8 | 968 | 144 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
