Assay Details
Gene Symbol: NFATC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014120434.1 | XP_013975909.1 | 289 | 105 | ||
XM_022408539.1 | XP_022264247.1 | 289 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
