Assay Details
Gene Symbol: POMK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_539949.6 | XP_539949.2 | 113 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_539949.6 transcripts.
Set Membership: |
![]() ![]() |
