Assay Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626920.3 | XP_005626977.1 | 2 - 3 | 684 | 93 | |
XM_014117917.2 | XP_013973392.1 | 2 - 3 | 885 | 93 | ||
XM_022424934.1 | XP_022280642.1 | 2 - 3 | 640 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() |
