Assay Details
Gene Symbol: ZNF483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626920.3 | XP_005626977.1 | 5 - 6 | 993 | 136 | |
XM_014117917.2 | XP_013973392.1 | 5 - 6 | 1194 | 136 | ||
XM_022424934.1 | XP_022280642.1 | 5 - 6 | 949 | 136 | ||
XM_022424936.1 | XP_022280644.1 | 4 - 5 | 1314 | 136 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
