Assay Details
Gene Symbol: RIPOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005640086.3 | XP_005640143.1 | 4 - 5 | 695 | 119 | |
XM_005640087.3 | XP_005640144.1 | 4 - 5 | 729 | 119 | ||
XM_005640088.3 | XP_005640145.1 | 4 - 5 | 571 | 119 | ||
XM_014110591.2 | XP_013966066.1 | 4 - 5 | 731 | 119 | ||
XM_545369.6 | XP_545369.4 | 4 - 5 | 730 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
