Assay Details
Gene Symbol: KIF1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168473.2 | NP_001161945.2 | 18 - 19 | 1740 | 58 | |
XM_005635949.3 | XP_005636006.1 | 20 - 21 | 1973 | 58 | ||
XM_005635951.3 | XP_005636008.1 | 20 - 21 | 1973 | 58 | ||
XM_005635952.3 | XP_005636009.1 | 19 - 20 | 1946 | 58 | ||
XM_005635953.3 | XP_005636010.1 | 19 - 20 | 1946 | 58 | ||
XM_014107680.2 | XP_013963155.1 | 20 - 21 | 2051 | 58 | ||
XM_014107681.2 | XP_013963156.1 | 20 - 21 | 2051 | 58 | ||
XM_014107683.2 | XP_013963158.1 | 20 - 21 | 2051 | 58 | ||
XM_014107688.2 | XP_013963163.1 | 19 - 20 | 1946 | 58 | ||
XM_022409568.1 | XP_022265276.1 | 20 - 21 | 2051 | 58 | ||
XM_022409569.1 | XP_022265277.1 | 20 - 21 | 2051 | 58 | ||
XM_022409570.1 | XP_022265278.1 | 20 - 21 | 1973 | 58 | ||
XM_022409571.1 | XP_022265279.1 | 19 - 20 | 1946 | 58 | ||
XM_022409572.1 | XP_022265280.1 | 19 - 20 | 1946 | 58 | ||
XM_022409573.1 | XP_022265281.1 | 20 - 21 | 2051 | 58 | ||
XM_022409574.1 | XP_022265282.1 | 20 - 21 | 2051 | 58 | ||
XM_022409575.1 | XP_022265283.1 | 20 - 21 | 1973 | 58 | ||
XM_022409576.1 | XP_022265284.1 | 19 - 20 | 1779 | 58 |
Gene Symbol: LOC100855783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107689.2 | XP_013963164.1 | 17 - 18 | 1824 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() |
