Assay Details
Gene Symbol: UGT3A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003638987.3 | XP_003639035.1 | 2 - 3 | 279 | 137 | |
XM_022417919.1 | XP_022273627.1 | 2 - 3 | 342 | 137 | ||
XM_022417920.1 | XP_022273628.1 | 2 - 3 | 226 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() |
