Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022419202.1 | XP_022274910.1 | 15 - 16 | 2009 | 51 | |
XM_022419203.1 | XP_022274911.1 | 15 - 16 | 2003 | 51 | ||
XM_022419204.1 | XP_022274912.1 | 15 - 16 | 1970 | 51 | ||
XM_022419205.1 | XP_022274913.1 | 14 - 15 | 1889 | 51 | ||
XM_022419206.1 | XP_022274914.1 | 15 - 16 | 2009 | 51 | ||
XM_022419207.1 | XP_022274915.1 | 15 - 16 | 2087 | 51 | ||
XM_022419208.1 | XP_022274916.1 | 14 - 15 | 2036 | 51 | ||
XM_022419210.1 | XP_022274918.1 | 9 - 10 | 1702 | 51 | ||
XM_022419211.1 | XP_022274919.1 | 10 - 11 | 1879 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
