Assay Details
Gene Symbol: C1QTNF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022422659.1 | XP_022278367.1 | 1682 | 80 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_022422659.1 transcripts.
Set Membership: |
![]() ![]() |
