Assay Details
Gene Symbol: TENT5D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_549103.5 | XP_549103.2 | 1 - 2 | 124 | 116 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_549103.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
