Assay Details
Gene Symbol: ERRFI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_546756.6 | XP_546756.1 | 140 | 112 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_546756.6 transcripts.
Set Membership: |
![]() ![]() |
