Assay Details
Gene Symbol: SMIM19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005629800.3 | XP_005629857.1 | 398 | 68 | ||
XM_005629801.3 | XP_005629858.1 | 514 | 68 | |||
XM_022403974.1 | XP_022259682.1 | 285 | 68 | |||
XM_843762.5 | XP_848855.1 | 397 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
