Assay Details
Gene Symbol: SH3BGRL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005641530.3 | XP_005641587.1 | 3 - 4 | 526 | 146 | |
XM_005641531.3 | XP_005641588.1 | 3 - 4 | 370 | 146 | ||
XM_022415749.1 | XP_022271457.1 | 3 - 4 | 337 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
