Assay Details
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547932.6 | XP_547932.2 | 3 - 4 | 321 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLIRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_547932.6 | XP_547932.2 | 3 - 4 | 321 | 96 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |