Assay Details
Gene Symbol: SPOUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005625275.3 | XP_005625332.1 | 4 - 5 | 444 | 62 | |
XM_846054.5 | XP_851147.2 | 4 - 5 | 444 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
