Assay Details
Gene Symbol: SELENOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114757.1 | NP_001108229.1 | 2 - 3 | 214 | 57 | |
GenBank mRNA | DN370248.1 | - | 2 - 3 | 211 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() |
