Assay Details
Gene Symbol: FAM174A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005618062.2 | XP_005618119.1 | 566 | 70 | ||
XM_845934.4 | XP_851027.2 | 553 | 70 |

More Information
Set Membership: |
![]() ![]() |
