Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115855.2 | XP_013971330.1 | 14 - 15 | 2688 | 150 | |
XM_014115856.2 | XP_013971331.1 | 14 - 15 | 2687 | 150 | ||
XM_014115857.2 | XP_013971332.1 | 13 - 14 | 2660 | 150 | ||
XM_014115858.2 | XP_013971333.1 | 13 - 14 | 2660 | 150 | ||
XM_014115859.2 | XP_013971334.1 | 14 - 15 | 2689 | 150 | ||
XM_014115860.2 | XP_013971335.1 | 9 - 10 | 770 | 150 | ||
XM_014115861.2 | XP_013971336.1 | 9 - 10 | 790 | 150 | ||
XM_014115862.2 | XP_013971337.1 | 8 - 9 | 763 | 150 | ||
XM_014115863.2 | XP_013971338.1 | 9 - 10 | 756 | 150 | ||
XM_014115864.2 | XP_013971339.1 | 9 - 10 | 657 | 150 | ||
XM_014115865.2 | XP_013971340.1 | 9 - 10 | 808 | 150 | ||
XM_022421957.1 | XP_022277665.1 | 8 - 9 | 763 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() |
