Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115856.2 | XP_013971331.1 | 16 - 17 | 2917 | 107 | |
XM_014115858.2 | XP_013971333.1 | 15 - 16 | 2890 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
