Assay Details
Gene Symbol: MIA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014115855.2 | XP_013971330.1 | 20 - 21 | 3201 | 139 | |
XM_014115856.2 | XP_013971331.1 | 19 - 20 | 3191 | 139 | ||
XM_014115857.2 | XP_013971332.1 | 19 - 20 | 3173 | 139 | ||
XM_014115858.2 | XP_013971333.1 | 18 - 19 | 3164 | 139 | ||
XM_014115859.2 | XP_013971334.1 | 20 - 21 | 3202 | 139 | ||
XM_014115860.2 | XP_013971335.1 | 15 - 16 | 1283 | 139 | ||
XM_014115861.2 | XP_013971336.1 | 15 - 16 | 1303 | 139 | ||
XM_014115862.2 | XP_013971337.1 | 14 - 15 | 1276 | 139 | ||
XM_014115863.2 | XP_013971338.1 | 15 - 16 | 1269 | 139 | ||
XM_014115864.2 | XP_013971339.1 | 15 - 16 | 1170 | 139 | ||
XM_014115865.2 | XP_013971340.1 | 15 - 16 | 1321 | 139 | ||
XM_022421957.1 | XP_022277665.1 | 14 - 15 | 1276 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
