Assay Details
Gene Symbol: DESI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022424369.1 | XP_022280077.1 | 2 - 3 | 341 | 76 | |
XM_022424370.1 | XP_022280078.1 | 2 - 3 | 341 | 76 | ||
XM_847031.5 | XP_852124.1 | 2 - 3 | 341 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
