Assay Details
Gene Symbol: IMPA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005623092.3 | XP_005623149.2 | 3 - 4 | 263 | 78 | |
XM_022421642.1 | XP_022277350.1 | 3 - 4 | 371 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
