Assay Details
Gene Symbol: LOC106557520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114467.2 | XP_013969942.1 | 3 - 4 | 790 | 130 | |
XM_022420019.1 | XP_022275727.1 | 3 - 4 | 794 | 130 | ||
XM_022420020.1 | XP_022275728.1 | 3 - 4 | 423 | 130 | ||
XM_022420021.1 | XP_022275729.1 | 3 - 4 | 801 | 130 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014114467.2, XM_022420019.1, XM_022420020.1, XM_022420021.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
