Assay Details
Gene Symbol: LOC106557520 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014114467.2 | XP_013969942.1 | 10 - 11 | 1375 | 67 | |
XM_022420019.1 | XP_022275727.1 | 10 - 11 | 1379 | 67 | ||
XM_022420020.1 | XP_022275728.1 | 10 - 11 | 1008 | 67 | ||
XM_022420021.1 | XP_022275729.1 | 10 - 11 | 1386 | 67 | ||
XM_022420022.1 | XP_022275730.1 | 9 - 10 | 1269 | 67 | ||
XM_022420023.1 | XP_022275731.1 | 9 - 10 | 1275 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
