Assay Details
Gene Symbol: SEPTIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005635975.3 | XP_005636032.2 | 5 - 6 | 505 | 128 | |
XM_846816.6 | XP_851909.2 | 5 - 6 | 459 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() |
