Assay Details
Gene Symbol: SPIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014119343.2 | XP_013974818.1 | 1 - 2 | 64 | 91 | |
XM_849901.5 | XP_854994.1 | 1 - 2 | 64 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014119343.2, XM_849901.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
