Assay Details
Gene Symbol: KIAA1614 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022420919.1 | XP_022276627.1 | 3519 | 82 | ||
XM_022420920.1 | XP_022276628.1 | 4112 | 82 | |||
XM_022420921.1 | XP_022276629.1 | 3411 | 82 | |||
XM_022420922.1 | XP_022276630.1 | 3519 | 82 | |||
XM_022420923.1 | XP_022276631.1 | 3272 | 82 | |||
XM_022420924.1 | XP_022276632.1 | 3525 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() |
