Assay Details
Gene Symbol: ERG28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014116088.2 | XP_013971563.1 | 1 - 2 | 361 | 111 | |
XM_849684.4 | XP_854777.2 | 1 - 2 | 353 | 111 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014116088.2, XM_849684.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
