Assay Details
Gene Symbol: BANF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_849683.5 | XP_854776.1 | 1 - 2 | 139 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_849683.5 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
