Assay Details
Gene Symbol: C26H12orf43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014107972.2 | XP_013963447.1 | 4 - 5 | 588 | 71 | |
XM_014107973.2 | XP_013963448.1 | 3 - 4 | 333 | 71 | ||
XM_849238.4 | XP_854331.1 | 3 - 4 | 333 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
