Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005626174.3 | XP_005626231.1 | 5 - 6 | 561 | 87 | |
XM_850290.5 | XP_855383.3 | 5 - 6 | 585 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
