Assay Details
Gene Symbol: SELENOW | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001115012.1 | NP_001108484.1 | 4 - 5 | 216 | 102 | |
GenBank mRNA | CN005642.1 | - | 4 - 5 | 239 | 102 | |
DN370714.1 | - | 4 - 5 | 213 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
