Assay Details
Gene Symbol: LOC102156482 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005628203.3 | XP_005628260.1 | 1 | 420 | 90 |
Gene Symbol: MSI2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_022423184.1 | XP_022278892.1 | 6 - 7 | 407 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
