Assay Details
Gene Symbol: S100A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005622818.3 | XP_005622875.1 | 2 - 3 | 209 | 68 | |
XM_850064.5 | XP_855157.1 | 3 - 4 | 196 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005622818.3, XM_850064.5 transcripts.
Set Membership: |
![]() ![]() ![]() |
